Please login to your online account to display your discounted pricing

5-[2-Chloro-4-(trifluoromethyl)phenyl]-2-furaldehyde, 95%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SEW06174DA

 View more versions of this product

Catalog No. SEW06174DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 274.62
Quantity 1g
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
CAS 306936-04-9
Chemical Name or Material 5-[2-Chloro-4-(trifluoromethyl)phenyl]-2-furaldehyde
MDL Number MFCD01571128
Packaging Amber glass bottle
Molecular Formula C12H6ClF3O2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 5-[2-Chloro-4-(trifluoromethyl)phenyl]-2-furaldehyde
CAS 306936-04-9
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H6ClF3O2
Formula Weight 274.62
MDL Number MFCD01571128
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents