missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Chloro-4-nitrophenylcarbamoyl)-3-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H54513MD

 View more versions of this product

Catalog No. AAH54513MD



Specifications

Specifications

5-(2-Chloro-4-nitrophenylcarbamoyl)-3-fluorobenzeneboronic acid
1449132-61-9
MFCD20126189
JGVQEEYFVVEKHQ-UHFFFAOYSA-N
[3-[(2-chloro-4-nitrophenyl)carbamoyl]-5-fluorophenyl]boronic acid
73995848
97%
250mg
C13H9BClFN2O5
3-2-chloro-4-nitrophenyl carbamoyl-5-fluorophenylboronic acid
B(C1=CC(=CC(=C1)F)C(=O)NC2=C(C=C(C=C2)[N+](=O)[O-])Cl)(O)O
338.482
338.49

Chemical Identifiers

1449132-61-9
338.482
JGVQEEYFVVEKHQ-UHFFFAOYSA-N
73995848
B(C1=CC(=CC(=C1)F)C(=O)NC2=C(C=C(C=C2)[N+](=O)[O-])Cl)(O)O
C13H9BClFN2O5
MFCD20126189
3-2-chloro-4-nitrophenyl carbamoyl-5-fluorophenylboronic acid
[3-[(2-chloro-4-nitrophenyl)carbamoyl]-5-fluorophenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents