Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Bromophenyl)-1H-tetrazole, 98+%

$45.72 - $732.70

Chemical Identifiers

Chemical Name or Material 5-(2-Bromophenyl)-1H-tetrazole
CAS 73096-42-1
Molecular Formula C7H5BrN4
Formula Weight 225.05
MDL Number MFCD00151799
Beilstein 148567
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2196603

 
alfa aesar™
B2196603
1g Each for $45.72

AAB2196606

 
alfa aesar™
B2196606
5g Each for $166.24

AAB2196614

 
alfa aesar™
B2196614
25g Each for $732.70
Description & Specifications

Specifications

Formula Weight 225.05
Chemical Name or Material 5-(2-Bromophenyl)-1H-tetrazole
Assay ≥98%
Molecular Formula C7H5BrN4
MDL Number MFCD00151799
CAS 73096-42-1
Melting Point 181° to 183°C
Beilstein 148567
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.