missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(2-Bromoethyl)-2,3-dihydrobenzofuran, TCI America™

$93.20 - $314.73

Chemical Identifiers

CAS 127264-14-6
Molecular Formula C10H11BrO
Molecular Weight (g/mol) 227.101
MDL Number MFCD06797641
InChI Key JRKZQRRYNCMSCB-UHFFFAOYSA-N
Synonym 5-(2-Bromoethyl)coumaran
PubChem CID 21831160
IUPAC Name 5-(2-bromoethyl)-2,3-dihydro-1-benzofuran
SMILES C1COC2=C1C=C(C=C2)CCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B40665G View Documents TCI America
B40665G
5g Each for $93.20
 
B406625G View Documents TCI America
B406625G
25g Each for $314.73
 
Specifications

Chemical Identifiers

127264-14-6
227.101
JRKZQRRYNCMSCB-UHFFFAOYSA-N
21831160
C1COC2=C1C=C(C=C2)CCBr
C10H11BrO
MFCD06797641
5-(2-Bromoethyl)coumaran
5-(2-bromoethyl)-2,3-dihydro-1-benzofuran

Specifications

Undesignated
5g
C10H11BrO
5-(2-Bromoethyl)coumaran
C1COC2=C1C=C(C=C2)CCBr
227.101
227.10
≥98.0% (GC)
67°C
127264-14-6
MFCD06797641
JRKZQRRYNCMSCB-UHFFFAOYSA-N
5-(2-bromoethyl)-2,3-dihydro-1-benzofuran
21831160
Crystal-Powder at 20°C
5-(2-Bromoethyl)-2,3-dihydrobenzofuran
Safety and Handling

Safety and Handling

TSCA : No