Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt, 97%

$69.27 - $181.64

Chemical Identifiers

Chemical Name or Material 5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt
CAS 100900-07-0
Molecular Formula C12H13N2NaO3S
Formula Weight 288.3
MDL Number MFCD00051474
Beilstein 4283850
Synonym EDANS sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0655503

  SDS
alfa aesar™
L0655503
1g Each for $181.64

AAL06555MD

  SDS
alfa aesar™
L06555MD
250mg Each for $69.27
Description & Specifications

Specifications

Formula Weight 288.3
Chemical Name or Material 5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt
MDL Number MFCD00051474
CAS 100900-07-0
Melting Point >300°C
Assay 97%
Molecular Formula C12H13N2NaO3S
Beilstein 4283850
Synonym EDANS sodium salt
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.