missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2,6-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H54222MD

 View more versions of this product

Catalog No. AAH54222MD



Specifications

Specifications

5-(2,6-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid
1449142-45-3
MFCD20040158
CHJDUXDVLGUAJJ-UHFFFAOYSA-N
[5-[(2,6-dimethylphenyl)carbamoyl]-2-fluorophenyl]boronic acid
73995698
97%
250mg
C15H15BFNO3
5-2,6-dimethylphenyl carbamoyl-2-fluorophenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC=C2C)C)F)(O)O
287.097
287.09

Chemical Identifiers

1449142-45-3
287.097
CHJDUXDVLGUAJJ-UHFFFAOYSA-N
73995698
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC=C2C)C)F)(O)O
C15H15BFNO3
MFCD20040158
5-2,6-dimethylphenyl carbamoyl-2-fluorophenylboronic acid
[5-[(2,6-dimethylphenyl)carbamoyl]-2-fluorophenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents