Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(2,5-Dichlorophenyl)-2-furoic acid, 97%

$37.36 - $146.46

Chemical Identifiers

Chemical Name or Material 5-(2,5-Dichlorophenyl)-2-furoic acid
CAS 186830-98-8
Molecular Formula C11H6Cl2O3
Formula Weight 257.07
MDL Number MFCD02213561
Synonym 5-(2,5-Dichlorophenyl)furan-2-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5199703

  SDS
alfa aesar™
H5199703
1g Each for $37.36

AAH5199706

  SDS
alfa aesar™
H5199706
5g Each for $146.46
Description & Specifications

Specifications

Formula Weight 257.07
Melting Point 233° to 237°C
MDL Number MFCD02213561
Molecular Formula C11H6Cl2O3
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
Quantity 1g
Synonym 5-(2,5-Dichlorophenyl)furan-2-carboxylic acid
CAS 186830-98-8
Assay 97%
Chemical Name or Material 5-(2,5-Dichlorophenyl)-2-furoic acid
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C