Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(1-Pyrrolidinylmethyl)thiophene-2-boronic acid pinacol ester, 97%

$75.03 - $287.80

Chemical Identifiers

Chemical Name or Material 5-(1-Pyrrolidinylmethyl)thiophene-2-boronic acid pinacol ester
CAS 1218790-45-4
Molecular Formula C15H24BNO2S
Formula Weight 293.23
MDL Number MFCD11113037
Synonym 5-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-thienyl]pyrrolidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5111803

  SDS
alfa aesar™
H5111803
1g Each for $75.03

AAH5111806

  SDS
alfa aesar™
H5111806
5g Each for $287.80
Description & Specifications

Specifications

Formula Weight 293.23
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Chemical Name or Material 5-(1-Pyrrolidinylmethyl)thiophene-2-boronic acid pinacol ester
Molecular Formula C15H24BNO2S
Synonym 5-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-thienyl]pyrrolidine
MDL Number MFCD11113037
Quantity 1g
CAS 1218790-45-4
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.