missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(1-Pyrrolidinylmethyl)thiophene-2-boronic acid pinacol ester, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5111803
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
5-(1-Pyrrolidinylmethyl)thiophene-2-boronic acid pinacol ester | |
C15H24BNO2S | |
1 g | |
VWOUUXTUYSWOCT-UHFFFAOYSA-N | |
1-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]pyrrolidine | |
46739746 | |
97% |
1218790-45-4 | |
MFCD11113037 | |
1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl pyrrolidine, 5-1-pyrrolidinylmethyl thiophene-2-boronic acid pinacol ester, 5-pyrrolidine methyl-2-thiopheneboronic acid pinacol ester, amtb126, 1-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophen-2-ylmethyl-pyrrolidine, 1-5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl pyrrolidine, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-thienyl pyrrolidine | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)CN3CCCC3 | |
293.232 | |
293.23 |
Chemical Identifiers
1218790-45-4 | |
293.232 | |
VWOUUXTUYSWOCT-UHFFFAOYSA-N | |
46739746 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)CN3CCCC3 |
C15H24BNO2S | |
MFCD11113037 | |
1-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl pyrrolidine, 5-1-pyrrolidinylmethyl thiophene-2-boronic acid pinacol ester, 5-pyrrolidine methyl-2-thiopheneboronic acid pinacol ester, amtb126, 1-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophen-2-ylmethyl-pyrrolidine, 1-5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophen-2-yl methyl pyrrolidine, 5-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-thienyl pyrrolidine | |
1-[[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methyl]pyrrolidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures