Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(1-Pyrrolidinyl)-2-furaldehyde, 98%

$114.51 - $396.28

Chemical Identifiers

Chemical Name or Material 5-(1-Pyrrolidinyl)-2-furaldehyde
CAS 84966-28-9
Molecular Formula C9H11NO2
Formula Weight 165.19
MDL Number MFCD02629619
Synonym 5-(1-Pyrrolidino)-2-furaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5031603

  SDS
alfa aesar™
H5031603
1g Each for $396.28

AAH50316MD

  SDS
alfa aesar™
H50316MD
250mg Each for $114.51
Description & Specifications

Specifications

Formula Weight 165.19
Melting Point 82° to 84°C
Assay 98%
Chemical Name or Material 5-(1-Pyrrolidinyl)-2-furaldehyde
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 84966-28-9
Health Hazard 1 Warning
Synonym 5-(1-Pyrrolidino)-2-furaldehyde
Quantity 1g
Molecular Formula C9H11NO2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD02629619
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.