Please login to your online account to display your discounted pricing

5-(1,4-Diazepan-1-yl)-3-phenyl-1,2,4-thiadiazole, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  AW00426DE

 View more versions of this product

Catalog No. AW00426DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 260.36
Quantity 5g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 5-(1,4-Diazepan-1-yl)-3-phenyl-1,2,4-thiadiazole
Assay Percent Range 97%
Molecular Formula C13H16N4S
MDL Number MFCD00729015
CAS 306934-71-4
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 5-(1,4-Diazepan-1-yl)-3-phenyl-1,2,4-thiadiazole
CAS 306934-71-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H16N4S
Formula Weight 260.36
MDL Number MFCD00729015
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.