Please login to your online account to display your discounted pricing

5-(1,3-Oxazol-5-yl)-2-thiophenesulfonyl chloride, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC14203DE

 View more versions of this product

Catalog No. CC14203DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 249.7
Quantity 5g
Assay 97%
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
CAS 321309-40-4
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 96.5
MDL Number MFCD02681961
Packaging Amber Glass Bottle
Molecular Formula C7H4ClNO3S2
Chemical Name or Material 5-(1, 3-Oxazol-5-yl)-2-thiophenesulfonyl chloride
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 5-(1, 3-Oxazol-5-yl)-2-thiophenesulfonyl chloride
CAS 321309-40-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4ClNO3S2
Formula Weight 249.7
MDL Number MFCD02681961
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents