missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(1,3-Oxazol-5-yl)-2-thiophenesulfonyl chloride, 97%, Maybridge™

Chemical Identifiers

CAS 321309-40-4
Molecular Formula C7H4ClNO3S2
Molecular Weight (g/mol) 249.683
MDL Number MFCD02681961
InChI Key MAGTZUMWQUDWAH-UHFFFAOYSA-N
Synonym 2-thiophenesulfonyl chloride, 5-5-oxazolyl, 2-thiophenesulfonyl chloride, 5-5-oxazolyl-9ci, 2-thiophenesulfonylchloride, 5-5-oxazolyl, 5-1,3-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-1,3-oxazol-5-yl thiophene-2-sulphonyl chloride, 5-1,3-oxazol-5-yl-2-thiophene sulfonyl chloride, 5-1,3-oxazol-5-yl-2-thiophenesulfonyl chloride, 5-oxazol-5-yl thiophene-2-sulfonyl chloride, chloro 5-1,3-oxazol-5-yl 2-thienyl sulfone
PubChem CID 2776411
IUPAC Name 5-(1,3-oxazol-5-yl)thiophene-2-sulfonyl chloride
SMILES C1=C(SC(=C1)S(=O)(=O)Cl)C2=CN=CO2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC14203DA
SDS View Documents
Fisher Scientific Company
CC14203DA
1g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC14203DE
SDS View Documents
Fisher Scientific Company
CC14203DE
5g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

321309-40-4
249.683
MAGTZUMWQUDWAH-UHFFFAOYSA-N
2776411
C1=C(SC(=C1)S(=O)(=O)Cl)C2=CN=CO2
C7H4ClNO3S2
MFCD02681961
2-thiophenesulfonyl chloride, 5-5-oxazolyl, 2-thiophenesulfonyl chloride, 5-5-oxazolyl-9ci, 2-thiophenesulfonylchloride, 5-5-oxazolyl, 5-1,3-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-1,3-oxazol-5-yl thiophene-2-sulphonyl chloride, 5-1,3-oxazol-5-yl-2-thiophene sulfonyl chloride, 5-1,3-oxazol-5-yl-2-thiophenesulfonyl chloride, 5-oxazol-5-yl thiophene-2-sulfonyl chloride, chloro 5-1,3-oxazol-5-yl 2-thienyl sulfone
5-(1,3-oxazol-5-yl)thiophene-2-sulfonyl chloride

Specifications

Amber Glass Bottle
321309-40-4
100.0
C7H4ClNO3S2
2-thiophenesulfonyl chloride, 5-5-oxazolyl, 2-thiophenesulfonyl chloride, 5-5-oxazolyl-9ci, 2-thiophenesulfonylchloride, 5-5-oxazolyl, 5-1,3-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-1,3-oxazol-5-yl thiophene-2-sulphonyl chloride, 5-1,3-oxazol-5-yl-2-thiophene sulfonyl chloride, 5-1,3-oxazol-5-yl-2-thiophenesulfonyl chloride, 5-oxazol-5-yl thiophene-2-sulfonyl chloride, chloro 5-1,3-oxazol-5-yl 2-thienyl sulfone
C1=C(SC(=C1)S(=O)(=O)Cl)C2=CN=CO2
249.683
249.7
5-(1, 3-Oxazol-5-yl)-2-thiophenesulfonyl chloride
1g
96.5
97%
MFCD02681961
MAGTZUMWQUDWAH-UHFFFAOYSA-N
5-(1,3-oxazol-5-yl)thiophene-2-sulfonyl chloride
2776411
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger