Please login to your online account to display your discounted pricing

5-[(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)methyl]-2-furaldehyde, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-[(1, 3-Dioxo-1, 3-dihydro-2H-isoindol-2-yl)methyl]-2-furaldehyde
CAS 116750-06-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H9NO4
Formula Weight 255.23
MDL Number MFCD02180376
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM10754CB

 
fisher scientific company
KM10754CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 255.23
Quantity 250mg
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 116750-06-2
Chemical Name or Material 5-[(1, 3-Dioxo-1, 3-dihydro-2H-isoindol-2-yl)methyl]-2-furaldehyde
Molecular Formula C14H9NO4
MDL Number MFCD02180376