Please login to your online account to display your discounted pricing

5-[(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)methyl]-2-furaldehyde, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM10754CB

 View more versions of this product

Catalog No. KM10754CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 255.23
Quantity 250mg
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 116750-06-2
Chemical Name or Material 5-[(1, 3-Dioxo-1, 3-dihydro-2H-isoindol-2-yl)methyl]-2-furaldehyde
Molecular Formula C14H9NO4
MDL Number MFCD02180376

Specifications

Chemical Name or Material 5-[(1, 3-Dioxo-1, 3-dihydro-2H-isoindol-2-yl)methyl]-2-furaldehyde
CAS 116750-06-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H9NO4
Formula Weight 255.23
MDL Number MFCD02180376