Please login to your online account to display your discounted pricing

(4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  337860010

 View more versions of this product

Catalog No. AC337860010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 229.35
Quantity 1g
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material (4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
MDL Number MFCD01862126
Molecular Formula C10H19NO3Si
CAS 82938-50-9
CAS Min % 98.5

Specifications

Chemical Name or Material (4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid
Name Note 99%
CAS 82938-50-9
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C10H19NO3Si
Formula Weight 229.35
MDL Number MFCD01862126