missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99%, ACROS Organics™
$510.86 - $510.86
Chemical Identifiers
CAS | 82938-50-9 |
---|---|
Molecular Formula | C10H19NO3Si |
Molecular Weight (g/mol) | 229.351 |
MDL Number | MFCD01862126 |
InChI Key | LIEWITJXZYCDLE-ZETCQYMHSA-N |
Synonym | 2s-1-tert-butyl dimethyl silyl-4-oxo-azetidine-2-carboxylic acid, 2s-1-tert-butyl dimethyl silyl-4-oxoazetidine-2-carboxylic acid, 2s-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid, 2s-4-oxo-1-1,1,2,2-tetramethyl-1-silapropyl azetidine-2-carboxylic acid, 4s-1-t-butyldimethylsilylazetidin-2-one-4-carboxylic acid, 4s-n-tert-butyldimethylsilyl azetidin-2-one-4-carboxylic acid, 4s-n-tert-butyldimethylsilyl azetidin-2-one-carboxylic acid, s-1-t-butyldimethylsilyl-4-oxo-2-azetidinecarboxylic acid, s-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid, s-n-tert-butyldimethylsilyl-4-oxo-azetidin-2 carboxylic acid |
PubChem CID | 10489602 |
IUPAC Name | (2S)-1-[tert-butyl(dimethyl)silyl]-4-oxoazetidine-2-carboxylic acid |
SMILES | CC(C)(C)[Si](C)(C)N1C(CC1=O)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC337862500
|
Thermo Fisher Scientific
337862500 |
250mg | Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC337860010
|
Thermo Fisher Scientific
337860010 |
1g | Glass bottle |
Each for $510.86
|
|
Chemical Identifiers
82938-50-9 | |
229.351 | |
LIEWITJXZYCDLE-ZETCQYMHSA-N | |
10489602 | |
CC(C)(C)[Si](C)(C)N1C(CC1=O)C(=O)O |
C10H19NO3Si | |
MFCD01862126 | |
2s-1-tert-butyl dimethyl silyl-4-oxo-azetidine-2-carboxylic acid, 2s-1-tert-butyl dimethyl silyl-4-oxoazetidine-2-carboxylic acid, 2s-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid, 2s-4-oxo-1-1,1,2,2-tetramethyl-1-silapropyl azetidine-2-carboxylic acid, 4s-1-t-butyldimethylsilylazetidin-2-one-4-carboxylic acid, 4s-n-tert-butyldimethylsilyl azetidin-2-one-4-carboxylic acid, 4s-n-tert-butyldimethylsilyl azetidin-2-one-carboxylic acid, s-1-t-butyldimethylsilyl-4-oxo-2-azetidinecarboxylic acid, s-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid, s-n-tert-butyldimethylsilyl-4-oxo-azetidin-2 carboxylic acid | |
(2S)-1-[tert-butyl(dimethyl)silyl]-4-oxoazetidine-2-carboxylic acid |
Specifications
Glass Bottle | |
250mg | |
98.5 | |
99% | |
MFCD01862126 | |
LIEWITJXZYCDLE-ZETCQYMHSA-N | |
(2S)-1-[tert-butyl(dimethyl)silyl]-4-oxoazetidine-2-carboxylic acid | |
10489602 | |
99% |
135.0°C to 137.0°C | |
82938-50-9 | |
100.0 | |
C10H19NO3Si | |
2s-1-tert-butyl dimethyl silyl-4-oxo-azetidine-2-carboxylic acid, 2s-1-tert-butyl dimethyl silyl-4-oxoazetidine-2-carboxylic acid, 2s-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid, 2s-4-oxo-1-1,1,2,2-tetramethyl-1-silapropyl azetidine-2-carboxylic acid, 4s-1-t-butyldimethylsilylazetidin-2-one-4-carboxylic acid, 4s-n-tert-butyldimethylsilyl azetidin-2-one-4-carboxylic acid, 4s-n-tert-butyldimethylsilyl azetidin-2-one-carboxylic acid, s-1-t-butyldimethylsilyl-4-oxo-2-azetidinecarboxylic acid, s-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid, s-n-tert-butyldimethylsilyl-4-oxo-azetidin-2 carboxylic acid | |
CC(C)(C)[Si](C)(C)N1C(CC1=O)C(=O)O | |
229.351 | |
229.35 | |
(4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99% |
Safety and Handling
TSCA : TSCA