Please login to your online account to display your discounted pricing

Alfa Aesar™ (4S)-(-)-Isopropyl-2-oxazolidinone, 98%

$81.36 - $310.39

Chemical Identifiers

Chemical Name or Material (4S)-(-)-Isopropyl-2-oxazolidinone
CAS 17016-83-0
Molecular Formula C6H11NO2
Formula Weight 129.16
MDL Number MFCD00010847
Beilstein 3588198
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1402903

 
alfa aesar™
A1402903
1g Each for $81.36

AAA1402906

 
alfa aesar™
A1402906
5g Each for $310.39
Description & Specifications

Specifications

Formula Weight 129.16
Chemical Name or Material (4S)-(-)-Isopropyl-2-oxazolidinone
Assay 98%
Molecular Formula C6H11NO2
MDL Number MFCD00010847
CAS 17016-83-0
Beilstein 3588198
Melting Point 70° to 74°C
Quantity 1g
Optical Rotation −17° (c=6 in Ethanol)