Please login to your online account to display your discounted pricing

Alfa Aesar™ (4S,5S)-(+)-4,5-Bis(fluoromethyl)-2,2-dimethyl-1,3-dioxolane, 90+%

$352.58 - $352.58

Chemical Identifiers

Chemical Name or Material (4S,5S)-(+)-4,5-Bis(fluoromethyl)-2,2-dimethyl-1,3-dioxolane
CAS 84409-70-1
Molecular Formula C7H12F2O2
Formula Weight 166.17
MDL Number MFCD01074597
Synonym (+)-2,3-O-Isopropylidiene-1,4-difluoro-2,3-butanediol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2776603

  SDS
alfa aesar™
H2776603
1g Each for $352.58
Description & Specifications

Specifications

Formula Weight 166.17
MDL Number MFCD01074597
Chemical Name or Material (4S,5S)-(+)-4,5-Bis(fluoromethyl)-2,2-dimethyl-1,3-dioxolane
CAS 84409-70-1
Quantity 1g
Synonym (+)-2,3-O-Isopropylidiene-1,4-difluoro-2,3-butanediol
Molecular Formula C7H12F2O2
Assay ≥90%