Please login to your online account to display your discounted pricing

(4S)-(-)-4-Isopropyl-2-oxazolidinone, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  297860050

 View more versions of this product

Catalog No. AC297860050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 129.16
Physical Form Crystalline Powder
Quantity 5g
Color White
Melting Point 70° to 74°C
Specific Rotation Condition −18° (20°C c=1,ethanol)
CAS Max % 100.0
Packaging Glass bottle
Molecular Formula C6H11NO2
Chemical Name or Material (4S)-(-)-4-Isopropyl-2-oxazolidinone
CAS 17016-83-0
Assay Percent Range 98%
CAS Min % 97.5
Infrared Spectrum Authentic
MDL Number MFCD00010847
Specific Rotation -16° to -20° (20°C, 589nm) (c=1, ethanol)

Specifications

Chemical Name or Material (4S)-(-)-4-Isopropyl-2-oxazolidinone
CAS 17016-83-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H11NO2
Formula Weight 129.16
MDL Number MFCD00010847