Please login to your online account to display your discounted pricing

(4S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  334840010

 View more versions of this product

Catalog No. AC334840010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 233.27
Quantity 1g
TSCA TSCA
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C13H15NO3
MDL Number MFCD00269688
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material (4S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone
Assay Percent Range 97%
CAS 101711-78-8
Packaging Glass bottle
Name Note 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (4S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone
Name Note 97%
CAS 101711-78-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C13H15NO3
Formula Weight 233.27
MDL Number MFCD00269688
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents