Please login to your online account to display your discounted pricing

(4S)-(-)-4,5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (4S)-(-)-4, 5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole
CAS 148461-14-7
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C24H24NOP
Formula Weight 373.43g/mol
MDL Number MFCD03427599
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC353731000

  SDS
fisher scientific acros
353731000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation -43° to -46° (20°C, 589nm) (C=1, CHCl3)
Color White
Quantity 100mg
Chemical Name or Material (4S)-(-)-4, 5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole
CAS 148461-14-7
CAS Min % 97.5
CAS Max % 100.0
Physical Form White crystalline powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C24H24NOP
Formula Weight 373.43g/mol
MDL Number MFCD03427599
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.