Please login to your online account to display your discounted pricing

(4S)-(-)-4,5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole, 98%, ACROS Organics™


Quantity 100mg
Packaging Glass bottle

Manufacturer: fisher scientific acros  35373-1000

 View more versions of this product

Catalog No. AC353731000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 373.43
Quantity 100mg
Color White
CAS Max % 100.0
Chemical Name or Material (4S)-(-)-4, 5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation -43° to -46° (20°C, 589nm) (C=1, CHCl3)
Assay Percent Range 98%
Molecular Formula C24H24NOP
MDL Number MFCD03427599
CAS 148461-14-7
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (4S)-(-)-4, 5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole
CAS 148461-14-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C24H24NOP
Formula Weight 373.43
MDL Number MFCD03427599
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents