Please login to your online account to display your discounted pricing

(4R)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99%, ACROS Organics™


Quantity 1g

Manufacturer: fisher scientific acros  337840010

 View more versions of this product

Catalog No. AC337840010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder or Chunks
Formula Weight 229.35
Quantity 1g
Color White to off-white
Melting Point 137° to 141°C
Chemical Name or Material (4R)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid
Assay Percent Range 99%
Molecular Formula C10H19NO3Si
CAS 162856-35-1
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (4R)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H19NO3Si
Formula Weight 229.35