Please login to your online account to display your discounted pricing

(4R)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (4R)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H19NO3Si
Formula Weight 229.35
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC337840010

  SDS
fisher scientific acros
337840010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC337840050

  SDS
thermo fisher scientific
AC337840050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder or Chunks
Formula Weight 229.35
Quantity 1g
Color White to off-white
Melting Point 137° to 141°C
Chemical Name or Material (4R)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid
Assay Percent Range 99%
Molecular Formula C10H19NO3Si
CAS 162856-35-1
Infrared Spectrum Authentic