Please login to your online account to display your discounted pricing

(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC337630250

 View more versions of this product

Catalog No. AC337630250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (HPLC)
Color Clear colorless to yellow-brown, may darken in storage
Physical Form Liquid
Formula Weight 177.2
Quantity 25g
Specific Rotation Condition + 162.00 (20.00°C c=2,CHCL3)
CAS Max % 100.0
Specific Rotation +165° to +175° (20°C, 589nm) (c=2, CHCl3)
CAS 77943-39-6
Chemical Name or Material (4R, 5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone
Assay Percent Range 99%
Molecular Formula C10H11NO2
CAS Min % 98.5
Infrared Spectrum Authentic
MDL Number MFCD00010845
Packaging Glass bottle

Specifications

Chemical Name or Material (4R, 5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone
CAS 77943-39-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C10H11NO2
Formula Weight 177.2
MDL Number MFCD00010845