missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 337630010

 View more versions of this product

Catalog No. AC337630010



Description

Description

Specifications

Specifications

(4R, 5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone
Glass bottle
+ 162
118°C to 121°C
77943-39-6
100.0
C10H11NO2
2-oxazolidinone, 4-methyl-5-phenyl-, 4r,5s, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r-cis, 4-methyl-5-phenyl-2-oxazolidinone 4r-cis, 4alpha-methyl-5alpha-phenyl-2-oxazolidinone, 4r,5s-+-4-methyl-5-phenyl-2-oxazolidinone, 4r,5s-4-methyl-5-phenyl-1,3-oxazolidin-2-one, 4r,5s-4-methyl-5-phenyl-2-oxazolidinone, 4r,5s-4-methyl-5-phenyloxazolidin-2-one, meph-2-oxazolidinone, pubchem6871
CC1C(OC(=O)N1)C2=CC=CC=C2
177.2
177.2
99%
Authentic
+ 162.00 (20.00°C c=2,CHCL3)
Brown-Yellow to Colorless
1g
98.5
98.5% min. (HPLC)
MFCD00010845
PPIBJOQGAJBQDF-VXNVDRBHSA-N
(4R,5S)-4-methyl-5-phenyl-1,3-oxazolidin-2-one
2733812
Liquid

Chemical Identifiers

77943-39-6
177.2
PPIBJOQGAJBQDF-VXNVDRBHSA-N
2733812
CC1C(OC(=O)N1)C2=CC=CC=C2
C10H11NO2
MFCD00010845
2-oxazolidinone, 4-methyl-5-phenyl-, 4r,5s, 2-oxazolidinone, 4-methyl-5-phenyl-, 4r-cis, 4-methyl-5-phenyl-2-oxazolidinone 4r-cis, 4alpha-methyl-5alpha-phenyl-2-oxazolidinone, 4r,5s-+-4-methyl-5-phenyl-2-oxazolidinone, 4r,5s-4-methyl-5-phenyl-1,3-oxazolidin-2-one, 4r,5s-4-methyl-5-phenyl-2-oxazolidinone, 4r,5s-4-methyl-5-phenyloxazolidin-2-one, meph-2-oxazolidinone, pubchem6871
(4R,5S)-4-methyl-5-phenyl-1,3-oxazolidin-2-one
SDS Documents