missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 5706-08-1 |
---|---|
Molecular Formula | C11H6OS |
Molecular Weight (g/mol) | 186.23 |
InChI Key | WSOJIQBCVMSHLB-UHFFFAOYSA-N |
PubChem CID | 12872762 |
IUPAC Name | indeno[1,2-b]thiophen-4-one |
SMILES | C1=CC=C2C(=C1)C3=C(C2=O)C=CS3 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
I1003200MG |
TCI America
I1003200MG |
200mg |
Each for $110.00
|
|
I10031G |
TCI America
I10031G |
1g |
Each for $325.42
|
|
Chemical Identifiers
5706-08-1 | |
186.23 | |
12872762 | |
C1=CC=C2C(=C1)C3=C(C2=O)C=CS3 |
C11H6OS | |
WSOJIQBCVMSHLB-UHFFFAOYSA-N | |
indeno[1,2-b]thiophen-4-one |
Specifications
Undesignated | |
200mg | |
C11H6OS | |
C1=CC=C2C(=C1)C3=C(C2=O)C=CS3 | |
186.23 | |
186.23 | |
≥98.0% (GC) |
101°C | |
5706-08-1 | |
WSOJIQBCVMSHLB-UHFFFAOYSA-N | |
indeno[1,2-b]thiophen-4-one | |
12872762 | |
Crystal-Powder at 20°C | |
4H-Indeno[1,2-b]thiophen-4-one |
Safety and Handling
TSCA : No