Please login to your online account to display your discounted pricing

4F 4PP oxalate, Tocris


Selective 5-HT2A antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS052350

 View more versions of this product

Catalog No. 052350

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 429.49
Assay Percent Range >99%
Chemical Name or Material 4F 4PP oxalate
Synonym 4-(4-Fluorobenzoyl)-1-(4-phenylbutyl)piperidine oxalate
Molecular Formula C22H26FNO·C2H2O4
CAS 144734-36-1
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material 4F 4PP oxalate
Assay Percent Range >99%
Molecular Formula C22H26FNO·C2H2O4
Formula Weight 429.49
Synonym 4-(4-Fluorobenzoyl)-1-(4-phenylbutyl)piperidine oxalate
Accessories

Store at Room Temperature