Please login to your online account to display your discounted pricing

4F 4PP oxalate, Tocris

Selective 5-HT2A antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material 4F 4PP oxalate
Assay Percent Range >99%
Molecular Formula C22H26FNO·C2H2O4
Formula Weight 429.49
Synonym 4-(4-Fluorobenzoyl)-1-(4-phenylbutyl)piperidine oxalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

052310

 
r & d systems
RDS052310
10mg Each for $177.37

052350

 
r & d systems
RDS052350
50mg Each for $746.48
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 429.49
Chemical Name or Material 4F 4PP oxalate
Assay Percent Range >99%
Molecular Formula C22H26FNO·C2H2O4
CAS 144734-36-1
Synonym 4-(4-Fluorobenzoyl)-1-(4-phenylbutyl)piperidine oxalate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature