Please login to your online account to display your discounted pricing

4EGI-1, Tocris™

Inhibitor of eIF4E:eIF4G interaction

$277.92 - $1,154.80

Chemical Identifiers

Chemical Name or Material 4EGI-1
Assay Percent Range >98%
Molecular Formula C18H12Cl2N4O4S
Formula Weight 451.28
Synonym α-[2-[4-(3, 4-Dichlorophenyl)-2-thiazolyl]hydrazinylidene]-2-nitro-benzenepropanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

480050

 
r & d systems
RDS480050
50mg Each for $1,154.80

480010

 
r & d systems
RDS480010
10mg Each for $277.92
Description & Specifications

Specifications

Formula Weight 451.28
Quantity 50mg
Assay Percent Range >98%
Molecular Formula C18H12Cl2N4O4S
Recommended Storage Store at -20°C
Synonym α-[2-[4-(3, 4-Dichlorophenyl)-2-thiazolyl]hydrazinylidene]-2-nitro-benzenepropanoic acid
Chemical Name or Material 4EGI-1
CAS 315706-13-9
Safety and Handling

Store at -20°C