Please login to your online account to display your discounted pricing

Alfa Aesar™ 4alpha-Phorbol 12-myristate 13-acetate, 99%

Negative control for phorbol ester activation of protein kinase C

Chemical Identifiers

Chemical Name or Material 4alpha-Phorbol 12-myristate 13-acetate
CAS 63597-44-4
Formula Weight 616.83
MDL Number MFCD00153860
Merck Index 14,7332
Synonym 4α-PMA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ61099LB0

  SDS
alfa aesar™
J61099LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ61099MCR

  SDS
alfa aesar™
J61099MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 616.83
Chemical Name or Material 4alpha-Phorbol 12-myristate 13-acetate
CAS 63597-44-4
MDL Number MFCD00153860
Assay 99%
Merck Index 14,7332
Storage Note 1 -30° to -10°C
Physical Form Powder or Film
Quantity 5mg
Solubility Information Soluble in DMSO, methanol or ethanol; Dilute stock solution into aqueous medium at final concentrations of ∽2 to 5μM
Sensitivity Light Sensitive
Color White
Synonym 4α-PMA

Applications
4alpha-Phorbol 12-myristate 13-acetate is a non-tumor promoting phorbol ester derivative that has been used as a negative control for PMA, a tumor promoter and activator of PKC. However, 4α-PMA has also been shown to be a reasonably potent activator of TRPV4 channels, making it a useful tool for structure-activity investigations via comparison to the very similar compound, 4α-PDD, which demonstrates 50 times more affinity for TRPV4. Furthermore, this compound has also been shown to increase PGHS-2 mRNA significantly in murine keratinocytes.

Solubility
Soluble in DMSO, methanol or ethanol. Dilute stock solution into aqueous medium at final concentrations of ~2-5µM

Notes
Light sensitive. Incompatible with oxidizing agents and heat.

Safety and Handling

-30° to -10°C