missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Spectrum™ Labs Spectra/Por™ 7 2000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
$456.91 - $456.91
Specifications
MWCO | 2 kDa |
---|---|
Quantity | 1 m Tubing/Kit |
Includes:
- 1meter of pre-treated tubing, a pair of closures and 5 opening picks
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Diameter (Metric) | Price | Quantity | ||||
08-700-202
|
Spectrum™
132108T |
4.6 mL/cm vol./length | 24 mm |
Each for $456.91
|
|
||||
08-700-201
|
Spectrum™
132107T |
1.1 mL/cm vol./length | 11.5 mm |
Each for $456.91
|
|
||||
Description
- Hydrophilic, symmetric porositiy
- Equilibrated in water
- Packaged wet in preservative solution of 0.05% sodium azide
- For small experiments or qualifying a membrane for a particular application
Specifications
2 kDa | |
1 m Tubing/Kit |