Please login to your online account to display your discounted pricing

NorLake™ Scientific Humidity And Temperature Stability Test Chambers w/Glass Doors

Advanced engineered design for optimal storage temperature environments

Specifications

Certifications/Compliance UL, C-UL
Humidity Display LED
Refrigerant R-134a
View More Specs
Products
Catalog Number Mfr. No. Capacity Electrical Requirements Price Quantity    

22-650-852

 
nor lake inc
NSRI803WSG5H
2269L (79.9 cu. ft.) 230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-850

 
nor lake inc
NSRI803WSG0H
2269L (79.9 cu. ft.) 115V, 1PH, 60Hz, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-851

  Accessories
nor lake inc
NSRI803WSG4H
2269L (79.9 cu. ft.) 208-230V, 1PH, 60Hz, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-849

 
nor lake inc
NSRI522WSG5H
1473L (51.9 cu. ft.) 230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-847

 
nor lake inc
NSRI522WSG4H
1473L (51.9 cu. ft.) 208-230V, 1PH, 60Hz, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-845

 
nor lake inc
NSRI331WSG5H
940L (33.1 cu.ft.) 230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-846

 
nor lake inc
NSRI522WSG0H
1473L (51.9 cu. ft.) 115V, 1PH, 60Hz, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-843

 
nor lake inc
NSRI331WSG0H
940L (33.1 cu.ft.) 115V, 1PH, 60Hz, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-844

  Accessories
nor lake inc
NSRI331WSG4H
940L (33.1 cu.ft.) 208-230V, 1PH, 60Hz, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-842

 
nor lake inc
NSRI241WSG5H
684L (24.1 cu. ft.) 230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-841

  Accessories
nor lake inc
NSRI241WSG4H
684L (24.1 cu. ft.) 208-230V, 1PH, 60Hz, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-840

 
nor lake inc
NSRI241WSG0H
684L (24.1 cu. ft.) 115V, 1PH, 60Hz, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Certifications/Compliance UL, C-UL
Humidity Display LED
Refrigerant R-134a