Mettler Toledo™ NewClassic ME Precision Balances, Internal Calibration
Efficient and convenient performance for daily weighing. Mettler Toledo NewClassic ME Precision Balances with Internal Calibration are designed to last, with solid metal base and reinforced body and are ideal for weighing under harsh conditions.
$1,709.79 - $1,889.99
Specifications
Electrical Requirements 100/240V, 50/60Hz
Display Type Backlit LCD
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Readability Model Stabilization Time Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Readability Model Stabilization Time Price Quantity  
01-913-923
View Documents
Mettler Toledo™
30029106
4.2kg 0.01 g ME4002 1 sec. Each for $1,709.79
01-913-922
View Documents
Mettler Toledo™
30029085
320g 0.001 g ME303 1.5 sec. Each for $1,889.99
Description
Description
  • Essential functionality plus ergonomic design
  • Fast and reliable results
  • Offers internal calibration
  • State-of-the-art technology ensures stable and accurate results
  • Overload protection up to 100kg
  • Front-feet leveling
  • Trace your data with date and time function to meet ISO/GLP documentation requirements
  • Intuitive interface with clear readout: large numbers and backlit display
  • 10 built-in applications
  • Offer protection against dust and splashing liquids in everyday operation, prolonging balance life
  • Rounded edges and smooth surfaces for easy cleaning
  • Low temperature drift effect: 3.0ppm/°C
Applications
Formulation, Totaling, Dynamic weighing, Piece counting, Density, Percent weighing, Check weighing, Statistics, Free factor
Specifications
Specifications
100/240V, 50/60Hz
Backlit LCD