missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Whatman™ Qualitative Filter Papers - Prepleated Grade 113V

Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses

$327.59 - $966.00

Specifications

Format Circle
Grade 113V
Particle Retention >30μm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
09-927-963 Whatman™
1213500
50cm Pack of 100 Each for $966.00
09-927-962 Whatman™
1213240
24cm Pack of 100 Each for $327.59
Specifications Description & Specifications

Specifications

Circle
113V
>30μm