Please login to your online account to display your discounted pricing

Boekel Scientific Open Source Bio™ (OSB) Transmitter: Calibration Reagents

For use with Open Source Bio (OSB) Transmitter.

$28.77 - $51.15

Specifications

For Use With (Equipment) Open Source Bio (OSB) Transmitter
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    

11-700-159

 
boekel scientific
BKL9510021
Conductivity Calibration Reagents Each for $38.36

11-700-157

 
boekel scientific
BKL9510020
pH Calibration Reagents Each for $51.15

11-700-158

 
boekel scientific
BKL9510011
DO Calibration Reagents Each for $28.77
Description & Specifications

Specifications

For Use With (Equipment) Open Source Bio (OSB) Transmitter

For use with Open Source Bio (OSB) Transmitter.