Excelta™ Lazer Line™ Medium Tapered Relieved Flush Cutter
Made of carbon tool steel with induction hardened cutting blades to RHC 63-65
$186.94 - $194.14
Specifications
Material Carbon tool steel
Grip Type Soft cushioned foam
Length (Metric) 133.3mm
Length (English) 5.25 in.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Cut Type Length (Metric) Tip Length (English) Price Quantity  
Catalog Number Mfr. No. Cut Type Length (Metric) Tip Length (English) Price Quantity  
18-100-961 Excelta Corporation
9148E
Optimum flush 0.03 x 0.08mm 5.25 in. Each for $194.14
18-100-959 Excelta Corporation
9146E
Semi-flush 0.25 x 0.3mm 5.25 in. Each for $186.94
18-100-960 Excelta Corporation
9147E
Maximum flush 0.08 x 0.13mm 5.25 in. Each for $190.54
Specifications Description & Specifications
Specifications
Carbon tool steel
Soft cushioned foam
133.3mm
5.25 in.