missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nor-Lake™ Scientific Temperature Stability Test Chamber

The NorLake™ Temperature Stability Test Chamber is advanced engineered for optimal storage temperature environments.

Specifications

Certifications/Compliance UL, C-UL
Display LED
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Electrical Requirements Height (Metric) Exterior Width (Metric) Exterior Price Quantity  
Catalog Number Mfr. No. Electrical Requirements Height (Metric) Exterior Width (Metric) Exterior Price Quantity  
22-650-826 Accessories Nor Lake Inc
NSRI803WSW4
208-230V, 1PH, 60Hz, 7.5A; NEMA plug: 6-15P; NEMA Receptacle: L-14-30R 204cm 209cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-650-827 Nor Lake Inc
NSRI803WSW5
230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module 204cm 209cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-650-823 Accessories Nor Lake Inc
NSRI522WSW4
208-230V, 1PH, 60Hz, 7.5A; NEMA plug: 6-15P; NEMA Receptacle: 6-15R 204cm 139cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-650-820 Accessories Nor Lake Inc
NSRI331WSW4
208-230V, 1PH, 60Hz, 7.5A; NEMA plug: 6-15P; NEMA Receptacle: 6-15R 224cm 80cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-650-822 Nor-Lake™ Scientific
NSRI522WSW0
115V, 1PH, 60Hz, 14.7A; NEMA plug: 5-20P; NEMA Receptacle: 5-20R 204cm 139cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-650-825 Nor Lake Inc
NSRI803WSW0
115V, 1PH, 60Hz, 14.7A; NEMA plug: 5-20P; NEMA Receptacle: 5-20R 204cm 209cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-650-824 Nor Lake Inc
NSRI522WSW5
230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module 204cm 139cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-650-819 Nor Lake Inc
NSRI331WSW0
115V, 1PH, 60Hz, 14.7A; NEMA plug: 5-20P; NEMA Receptacle: 5-20R 224cm 80cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-650-817 Accessories Nor Lake Inc
NSRI241WSW4
208-230V, 1PH, 60Hz, 7.5A; NEMA plug: 6-15P; NEMA Receptacle: 6-15R 204cm 69cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-650-816 Nor Lake Inc
NSRI241WSW0
115V, 1PH, 60Hz, 14.7A; NEMA plug: 5-20P; NEMA Receptacle: 5-20R 204cm 69cm N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Certifications

UL listed, C-UL listed

Description & Specifications

Specifications

UL, C-UL
LED