Please login to your online account to display your discounted pricing

Nor-Lake™ Scientific Humidity And Temperature Stability Test Chamber

The NorLake™ Humidity And Temperature Stability Test Chamber is advanced engineered for optimal storage temperature environments.

Specifications

Certifications/Compliance UL, C-UL
Humidity Display LED
Humidity Range Solid door model: +4°C to +70°C (+39.2°F to +158°F); Glass door model: +20°C to +60°C (68°F to 140°F)
View More Specs
Products
Catalog Number Mfr. No. Depth (Metric) Exterior Electrical Requirements Height (Metric) Exterior Width (Metric) Exterior Price Quantity    

22-650-802

  Accessories
nor-lake™ scientific
NSRI241WSW0H
88cm 115V, 1PH, 60Hz, 14.7A; NEMA plug: 5-20P; NEMA Receptacle: 5-20R 204cm 69cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-812

  Accessories
nor lake inc
NSRI803WSW4H
88cm 208-230V, 1PH, 60Hz, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R 202cm 206cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-813

 
nor lake inc
NSRI803WSW5H
88cm 230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module 202cm 88cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-808

 
nor lake inc
NSRI522WSW0H
88cm 115V, 1PH, 60HZ, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R 204cm 139cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-809

  Accessories
nor-lake™ scientific
NSRI522WSW4H
88cm 208-230V, 1PH, 60HZ, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R 204cm 139cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-810

 
nor lake inc
NSRI522WSW5H
88cm 230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module 204cm 139cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-811

 
nor-lake™ scientific
NSRI803WSW0H
88cm 115V, 1PH, 60Hz, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R 204cm 209cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-806

  Accessories
nor lake inc
NSRI331WSW4H
91cm 208-230V, 1PH, 60HZ, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R 224cm 80cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-805

 
nor-lake™ scientific
NSRI331WSW0H
91cm 115V, 1PH, 60HZ, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R 224cm 78cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-803

  Accessories
nor-lake™ scientific
NSRI241WSW4H
88cm 208-230V, 1PH, 60HZ, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R 204cm 69cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Certifications/Compliance UL, C-UL
Humidity Display LED
Humidity Range Solid door model: +4°C to +70°C (+39.2°F to +158°F); Glass door model: +20°C to +60°C (68°F to 140°F)

UL listed, C-UL listed