Please login to your online account to display your discounted pricing

Brady™ Vinyl Tapes (B-595) for BMP71 Printer - Red

Indoor/Outdoor vinyl grade conforms to irregular, smooth or rough surfaces

$116.84 - $116.84

Specifications

For Use With (Application) Labeling pipes, walls, equipment, windows
For Use With (Equipment) BMP71 Printer
Material Vinyl
Product Type Vinyl Tape
Quantity 1/ea
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity    

17-111-051

 
brady™
M71C2000595RD**
15.24m x 5.08cm (50 ft. x 2 in.) Each for $116.84
Description & Specifications

Specifications

For Use With (Application) Labeling pipes, walls, equipment, windows
For Use With (Equipment) BMP71 Printer
Material Vinyl
Product Type Vinyl Tape
Quantity 1/ea
Color Red