Brady™ UXT Xtra Tough Universal Heavy Rolls
Made to handle virtually any liquid (water, petroleum and chemical-based)
$113.85 - $113.85
Specifications
For Use With (Application) Food Processing, High Tech Industries, Machining of small parts, Paint room, Pharmaceutical, Printing
Product Type UXT Xtra Tough Universal Heavy Roll
Packaging Quantity 1/Cs.
Absorbancy Heavy
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (English) Sorbing Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Capacity (English) Sorbing Dimensions (L x W) Price Quantity  
29-635-3227 View Documents Brady™
UXT15P
17 gal./bale 45.72m x 38.1cm (150 ft. x 15 in.) Pack for $113.85
Description
Description
  • Three-ply construction
  • Double-sided, low-linting coverstock and meltblown core
  • Abrasion resistant and non-abrasive performance
  • Perforated
Specifications
Specifications
Food Processing, High Tech Industries, Machining of small parts, Paint room, Pharmaceutical, Printing
UXT Xtra Tough Universal Heavy Roll
1/Cs.
Heavy