Please login to your online account to display your discounted pricing

Labconco™ Protector™ Echo™ Filtered Benchtop Fume Hoods: 8 ft. Width

Adaptable fully-featured fume hood with Neutrodine™ filtration technology

Specifications

Depth (Metric) 95.8cm
Height Overall (Metric) 167.6cm
Hertz 50/60Hz
Lighting Fluorescent
Width (Metric) 243.8cm
View More Specs
Products
Catalog Number Mfr. No. Amperage Includes Voltage Weight (Metric) Price Quantity    

13-118-363

 
labconco™
LCN182810002
10Amp Back, side windows, integral blower, 2 service fixtures, 2 electrical duplexes 100-115V 469.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-362

 
labconco™
LCN182810001
10Amp Back and side windows, integral blower, 2 service fixtures 100-115V 469.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-361

 
labconco™
LCN182810000
10Amp Back and side windows, integral blower 100-115V 464.9kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-360

 
labconco™
LCN181810002
10Amp Side windows, integral blower, 2 service fixtures, 2 electrical duplexes 100-115V 464.9kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-359

 
labconco™
LCN181810001
10Amp Side windows, integral blower, 2 service fixtures 100-115V 464.9kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-358

 
labconco™
LCN181810000
10Amp Side windows, integral blower 100-115V 460.4kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-355

 
labconco™
LCN180810000
10Amp Integral blower 100-115V 446.8kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-356

 
labconco™
LCN180810001
10Amp Integral blower, 2 service fixtures 100-115V 446.8kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-357

 
labconco™
LCN180810002
10Amp Integral blower, 2 service fixtures, 2 electrical duplexes 100-115V 451.3kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Depth (Metric) 95.8cm
Height Overall (Metric) 167.6cm
Hertz 50/60Hz
Lighting Fluorescent
Width (Metric) 243.8cm

AFNOR NF-X, ANSI Z9.5, ASHRAE 110, ASTM E84, CAN/CSA C22.2, SEFA 8 (Cabinet Surface Finish), SEFA 9, UL 61010