Please login to your online account to display your discounted pricing

Labconco™ Protector™ Echo™ Filtered Benchtop Fume Hoods: 6 ft. Width

Adaptable fully-featured fume hood with Neutrodine™ filtration technology

Specifications

Depth (Metric) 95.8cm
Height Overall (Metric) 167.6cm
Hertz 50/60Hz
Lighting Fluorescent
Width (Metric) 182.9cm
View More Specs
Products
Catalog Number Mfr. No. Amperage Includes Voltage Weight (Metric) Price Quantity    

13-118-352

 
labconco™
LCN182610020
5Amp Back and side windows, integral blower 208-230V 369.7kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-353

 
labconco™
LCN182610021
5Amp Back and side windows, integral blower; 2 service fixtures 208-230V 374.2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-350

 
labconco™
LCN181610021
5Amp Side windows, integral blower; 2 service fixtures 208-230V 369.7kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-349

 
labconco™
LCN181610020
5Amp Side windows, integral blower 208-230V 365.1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-346

 
labconco™
LCN180610020
5Amp Integral blower 208-230V 337.9kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-347

 
labconco™
LCN180610021
5Amp Integral blower, 2 service fixtures 208-230V 342.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-345

 
labconco™
LCN182610002
10Amp Back and side windows, integral blower, 2 service fixtures, 1 electrical duplex 100-115V 374.2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-343

 
labconco™
LCN182610000
10Amp Back and side windows, integral blower 100-115V 369.7kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-344

 
labconco™
LCN182610001
10Amp Back and side windows, integral blower, 2 service fixtures 100-115V 374.2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-342

 
labconco™
LCN181610002
10Amp Side windows, integral blower, 2 service fixtures, 1 electrical duplex 100-115V 369.7kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-341

 
labconco™
LCN181610001
10Amp Side windows, integral blower, 2 service fixtures 100-115V 369.7kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-339

 
labconco™
LCN180610002
10Amp Integral blower, 2 service fixtures, 1 electrical duplex 100-115V 342.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-340

 
labconco™
LCN181610000
10Amp Side windows, integral blower 100-115V 365.1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-338

 
labconco™
LCN180610001
10Amp Integral blower, 2 service fixtures 100-115V 342.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-337

 
labconco™
LCN180610000
10Amp Integral blower 100-115V 337.9kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Depth (Metric) 95.8cm
Height Overall (Metric) 167.6cm
Hertz 50/60Hz
Lighting Fluorescent
Width (Metric) 182.9cm

AFNOR NF-X, ANSI Z9.5, ASHRAE 110, ASTM E84, CAN/CSA C22.2, SEFA 8 (Cabinet Surface Finish), SEFA 9, UL 61010