Please login to your online account to display your discounted pricing

Labconco™ Protector™ Echo™ Filtered Benchtop Fume Hoods: 5 ft. Width

Adaptable fully-featured fume hood with Neutrodine™ filtration technology

Specifications

Depth (Metric) 95.8cm
Height Overall (Metric) 167.6cm
Hertz 50/60Hz
Lighting Fluorescent
Width (Metric) 152.4cm
View More Specs
Products
Catalog Number Mfr. No. Amperage Includes Voltage Weight (Metric) Price Quantity    

13-118-332

 
labconco™
LCN181510021
5Amp Side windows, integral blower; 2 service fixtures 208-230V 315.2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-335

 
labconco™
LCN182510021
5Amp Back and side windows, integral blower; 2 service fixtures 208-230V 319.8kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-327

 
labconco™
LCN182510002
10Amp Back and side windows, integral blower, 2 service fixtures, 1 electrical duplex 100-115V 319.8kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-331

 
labconco™
LCN181510020
5Amp Side windows, integral blower 208-230V 310.7kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-334

 
labconco™
LCN182510020
5Amp Back and side windows, integral blower 208-230V 315.2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-328

 
labconco™
LCN180510020
5Amp Integral blower 208-230V 297.1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-329

 
labconco™
LCN180510021
5Amp Integral blower, 2 service fixtures 208-230V 301.6kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-326

 
labconco™
LCN182510001
10Amp Back and side windows, integral blower, 2 service fixtures 100-115V 319.8kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-324

 
labconco™
LCN181510002
10Amp Side windows, integral blower, 2 service fixtures, 1 electrical duplex 100-115V 315.2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-325

 
labconco™
LCN182510000
10Amp Back and side windows, integral blower 100-115V 315.2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-322

 
labconco™
LCN181510000
10Amp Side windows, integral blower 100-115V 310.7kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-323

 
labconco™
LCN181510001
10Amp Side windows, integral blower, 2 service fixtures 100-115V 315.2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-319

 
labconco™
LCN180510000
10Amp Integral blower 100-115V 297.1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-320

 
labconco™
LCN180510001
10Amp Integral blower, 2 service fixtures 100-115V 301.6kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

13-118-321

 
labconco™
LCN180510002
10Amp Integral blower, 2 service fixtures, 1 electrical duplex 100-115V 301.6kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Depth (Metric) 95.8cm
Height Overall (Metric) 167.6cm
Hertz 50/60Hz
Lighting Fluorescent
Width (Metric) 152.4cm

AFNOR NF-X, ANSI Z9.5, ASHRAE 110, ASTM E84, CAN/CSA C22.2, SEFA 8 (Cabinet Surface Finish), SEFA 9, UL 61010