Please login to your online account to display your discounted pricing
BrandTech™ BRANDplates™ Insert Strips With Inlet Opening System
Available with two membranes PC or PET membrane
$1,476.76 - $1,476.76
Specifications
Type Insert Strips With Inlet Opening System
Sterile Yes
Surface Coating cellGrade™ plus
Quantity 12/Pk.
View More Specs
Products
Catalog Number Mfr. No. Membrane Pore Size Price Quantity    
14-380-850
brandtech™
782841
PC membrane 3μm Pack of 12 Each for $1,476.76
14-380-862
brandtech™
782871
PET membrane 8μm Pack of 12 Each for $1,476.76
14-380-838
brandtech™
782811
PET membrane 0.4μm Pack of 12 Each for $1,476.76
14-380-842
brandtech™
782821
PC membrane 1μm Pack of 12 Each for $1,476.76
14-380-858
brandtech™
782861
PC membrane 8μm Pack of 12 Each for $1,476.76
14-380-834
brandtech™
782801
PC membrane 0.4μm Pack of 12 Each for $1,476.76
Description & Specifications
Specifications
Type Insert Strips With Inlet Opening System
Sterile Yes
Surface Coating cellGrade™ plus
Quantity 12/Pk.