Fisherbrand™ Upright Set Leg Mounted Single Sided Slots With Power Plug
Snap-on accessories. No tools required
$271.72 - $286.85
Specifications
Adjustable Yes
Finish Painted steel
For Use With (Equipment) Workbench or table
Quantity 1 Ea.
Product Type Single Sided Slot with Power Plug
View More Specs
Products 2
Catalog Number Mfr. No. Height (Metric) Price Quantity  
Catalog Number Mfr. No. Height (Metric) Price Quantity  
25-100-353
View Documents Promotion Details
Fisherbrand™
UP48L
122 cm
Each for $286.85
25-100-351
View Documents Promotion Details
Fisherbrand™
UP36L
91.4 cm
Each for $271.72
Description
Description
 Easy snap on accessory with no tools required for assembly.
  • Heavy 5.08cm (2 in.) square frame
  • Uprights are bolted directly into leg
  • Snap-on accessories. No tools required
  • Required for adjustable shelves, bin box rails and lights
  • Powerplug and cord are mounted on one upright
  • Level shelves and accessories without measuring
Specifications
Specifications
Yes
Workbench or table
Single Sided Slot with Power Plug
Painted steel
1 Ea.
White, Black
SDS
missing translation for 'documents'
missing translation for 'documents'