missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell™ Blue 1H Half Mask Respirators
Designed with healthcare workers for healthcare workers
$45.21 - $78.00
Specifications
Suspension Type | Cradle |
---|---|
Disposable | No |
Includes | P100 filters and splash guards |
Material | Silicone |
For Use With (Application) | Healthcare professionals in environments ranging from the ER, OR, and IC to clinics and other medical facilities |
Includes: One pair of P100 particulate filters and filter splash guards.
Catalog Number | Mfr. No. | Color | Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Size | Price | Quantity | ||||
19-066-243
|
Honeywell™
B230070 |
Light Green | Large |
Each for $78.00
|
|
19-066-250
|
Honeywell™
B230080 |
Lilac | Large |
Each for $67.84
|
|
19-066-249
|
Honeywell™
B220080 |
Lilac | Medium |
Each for $67.84
|
|
19-066-242
|
Honeywell™
B220070 |
Light Green | Medium |
Each for $67.84
|
|
19-066-248
|
Honeywell™
B210080 |
Lilac | Small |
Each for $78.00
|
|
19-066-241
|
Honeywell™
B210070 |
Light Green | Small |
Each for $45.21
|
|
Description
Reusable respirator to protect healthcare workers from the hazards and challenges faced each day in infectious disease environments or during a pandemic
- Includes a speech diaphragm for better worker-to-patient speech clarity for improved communication
- Splash guards for the P100 filters help prevent blood borne pathogens from contaminating the exterior of the filters
- Splash guard over the exhalation port directs healthcare worker's exhaled breath away from patients
- Silicone facepiece is soft and flexible to provide workers with a comfortable fit for hours of use
- Dual sealing flange provides excellent facepiece seal
- Cradle suspension provides excellent support for a secure fit
- NIOSH approved (with P100 particulate filters only)
Order Info
Available in light green and lilac colors of scrubs.
Specifications
Cradle | |
P100 filters and splash guards | |
Healthcare professionals in environments ranging from the ER, OR, and IC to clinics and other medical facilities | |
Half-face respirator |
No | |
Silicone | |
Half mask |