Please login to your online account to display your discounted pricing
Whatman™ FTA™ Nucleic Acid Collection, Storage and Purification Cards: FTA PlantSaver
Collect, store, and transport DNA at room temperature
$423.14 - $423.14
Specifications
Product Type FTA PlantSaver card
For Use With (Application) Forensics, transgenic identification, transfusion medicine/HLA typing, plasmid screening, food and agriculture testing, drug discovery, genomics, STR analysis, animal identification, whole genome amplification
Format Card
View More Specs
Products
Catalog Number Mfr. No. Formulation Includes Quantity Price Quantity    
09-927-972
whatman™
WHTWB120068
Non-indicating Pack for $423.14
Description & Specifications
Specifications
Product Type FTA PlantSaver card
For Use With (Application) Forensics, transgenic identification, transfusion medicine/HLA typing, plasmid screening, food and agriculture testing, drug discovery, genomics, STR analysis, animal identification, whole genome amplification
Format Card