HyClone™ HyCell™ TransFx™-H Media
$150.27 - $150.27
Products 1
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
SH3093902 View Documents HyClone™
SH3093902
Each for $150.27
Description
Description
HyCell TransFx-H media for transient transfection of HEK293 cells and production of a variety of recombinant proteins manufactured under cGMP in ISO-compliant facilities.
Highlights:
  • Defined hydrolysate-free ADCF cell culture media formulation
  • Supports transient transfection and growth of multiple HEK293 cell lines
  • Supports the transient expression of a variety of recombinant proteins
  • Same media for transient transfection and production
  • Developed through our Metabolic Pathway Design process for high transfection efficiency, high viable cell yields, and recombinant protein production
  • Component traceability
  • Manufactured under cGMP in ISO-compliant facility
Order Info
Available in liquid form in 1L bottles, 5 to 500L bioprocess containers or custom packaging. The media is also available as a powder in pack sizes from 5 to 1,000L bioprocess production applications.